Learn More
Sino Biological Influenza A H5N1 (A/Anhui/1/2005) Hemagglutinin Protein (HA1 Subunit)
Supplier:  Sino Biological 11048VNAH2100
Encompass_Preferred
A DNA sequence encoding the the N-terminal segment (Met 1-Pro 337) of Influenza A virus H5N1 hemagglutinin (A/Anhui/1/2005 (H5N1)) (ABD 28180.1), termed as HA1, was expressed.
Catalog No. 50-161-6095