Learn More
Sino Biological Recombinant Human Beta-NGF Protein, HPLC-verified
Supplier:  Sino Biological 11050HNAC100
Encompass_Preferred
A DNA sequence encoding the mature form of human β-NGF (NP_002497.2) (Ser 122-Arg 239) was expressed.
Catalog No. 50-161-6101