Learn More
Sino Biological Recombinant Human TIM-1/HAVCR1 Protein (ECD, His Tag)
Supplier:  Sino Biological 11051H07H100
Encompass_Preferred
A DNA sequence encoding the mature form of human KIM1 extracellular domain (AAC39862.1) (Ser 21-Gly 290) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6104