Learn More
Sino Biological Influenza A H1N1 (A/Brisbane/59/2007) Hemagglutinin / HA Protein (ECD, His Tag)
Supplier:  Sino Biological 11052V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of Influenza A virus (A/Brisbane/59/2007 (H1N1)) (ACA28844.1) hemagglutinin (Met 1-Gln 528) (Native, HA1+HA2, uncleaved) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-6113