Learn More
Sino Biological Influenza A H1N1 (A/Brisbane/59/2007) Hemagglutinin Protein (HA1 Subunit) (His Tag)
Supplier:  Sino Biological 11052V08H1100
Encompass_Preferred
A DNA sequence encoding an N-terminal segment (Met 1-Arg 343) of Influenza A virus (A/Brisbane/59/2007 (H1N1)) (ACA28844.1) HA, termed as HA1 subunit, was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6115