Learn More
Sino Biological Influenza B (B/Florida/4/2006) Hemagglutinin Protein (HA2 Subunit) (Fc Tag)
Supplier:  Sino Biological 11053V04H2100
Encompass_Preferred
A DNA sequence encoding the C-terminal fragment (Gly 362-Ala 555) of influenza B virus (B/Florida/4/2006) (ACA33493.1) hemagglutinin, termed as HA2, was fused with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-161-6120