Learn More
Sino Biological Influenza B (B/Florida/4/2006) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11053V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of Influenza B (B/Florida/4/2006)) (ACA33493.1) hemagglutinin (Met 1-Ala 555) (Native, HA1+HA2, uncleaved) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-6123