Learn More
Sino Biological Influenza A H1N1 (A/California/04/2009) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11055V08H2100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of Influenza A virus (A/California/04/2009 (H1N1)) hemagglutinin (ACP41105.1) (Met 1-Gln 529) with cleavage site mutated (IQS→RQRRRRK, HA1+HA2, partially cleaved) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-6140