Learn More
Sino Biological Influenza A H3N2 (A/Brisbane/10/2007) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11056V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of Influenza A virus (A/Brisbane/10/2007 (H3N2)) (ABW23353.1) hemagglutinin (Met 1-Ile 531) (Native, HA1+HA2, uncleaved) was expressed with a C-terminal polyhistidine.
Catalog No. 50-161-6152