Learn More
Sino Biological Recombinant Cynomolgus IL-13 Protein (His Tag)
Supplier:  Sino Biological 11057C07H100
Encompass_Preferred
A DNA sequence encoding the mature form of Cynomolgus (Macaca fascicularis) IL-13 (ABG75889.1) (Ser 21-Asn 132) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6157