Learn More
Sino Biological Recombinant Cynomolgus IL-13 Protein
Supplier:  Sino Biological 11057CNAH1
Encompass_Preferred
A DNA sequence encoding the Cynomolgus (Macaca fascicularis) IL13 (ABG75889.1) (Met 1-Asn 132) precursor was expressed.
Catalog No. 50-161-6160