Learn More
Sino Biological Influenza A H1N1 (A/California/04/2009) Neuraminidase / NA (Fc Tag)
Supplier:  Sino Biological 11058V01H100
Encompass_Preferred
A DNA sequence encoding the mature form of Influenza A virus H1N1 (A/California/04/2009) neuraminidase (ACP41107.1) (His 36-Lys 469) was expressed with the fused Fc region of human IgG1 at the N-terminus linked by a peptide linker.
Catalog No. 50-161-6166