Learn More
Sino Biological Influenza A H5N1 (A/bar-headed goose/Qinghai/14/2008) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11059V08H2100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of Influenza A virus (A/bar-headed goose/Qinghai/14/2008 (H5N1)) (ACL28277.1) HA (Met 1-Gln 531) (Native, HA1+HA2, cleaved) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-6181