Learn More
Sino Biological Influenza A H5N1 (A/turkey/Turkey/1/2005) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11061V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/turkey/Turkey/1/2005(H5N1)) hemagglutinin (Q207Z6, with aa 339-342 mutated from "GERR" to "RESR") (Met1-Gln531), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6193