Learn More
Sino Biological Influenza A H5N1 (A/Vietnam/1194/2004) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11062V08H1100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of Influenza A virus (A/VietNam/1194/2004 (H5N1)) (AAT73273.1) hemagglutinin (Met 1-Gln 531) with cleavage site mutated (RERRRKKR →TETR, HA1+HA2, uncleaved) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-6204