Learn More
Sino Biological Recombinant Human LIMPII/SR-B2 Protein (His & hFc Tag)
Supplier:  Sino Biological 11063H03H100
Encompass_Preferred
A DNA sequence encoding the human SCARB2 (NP_005497.1) extracellular domain (Arg 27-Thr 432) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-6209