Learn More
Sino Biological Recombinant Human Podoplanin Protein (ECD, His & hFc Tag), HPLC-verified
Supplier:  Sino Biological 11065H03H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human PDPN (Q86YL7-1) (Met1-Lys123) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-6213