Learn More
Sino Biological VEGF165 Protein, Human, Cynomolgus, Recombinant, HPLC-verified
Supplier:  Sino Biological 11066HNAH100
Encompass_Preferred
A DNA sequence encoding the human / cynomolgus VEGF165 isoform (P15692-4) (Met1-Arg191) was expressed. Human and Cynomolgus VEGF165 sequences are identical.
Catalog No. 50-161-6438