Learn More
Sino Biological Influenza A H1N1 (A/Brevig Mission/1/1918) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11068V08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of Influenza A virus H1N1 (A/Brevig Mission/1/1918 (H1N1)) hemagglutinin (AAD17229.1) (Met 1-Gln 529) (Native, HA1+HA2, uncleaved) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-6443