Learn More
Sino Biological Human respiratory syncytial virus (RSV) (A, rsb1734) glycoprotein G / RSV-G Protein (95% Homology) (His Tag)
Supplier:  Sino Biological 11070V08H100
Encompass_Preferred
A DNA sequence encoding the glycoprotein G extracellular domain (Asn 66-Arg 297) of human respiratory syncytial virus A (95 % homologous with strain rsb1734) (P27022-1) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-6457