Learn More
Sino Biological Recombinant Human TrkA Protein (His & hFc Tag), HPLC-verified
Supplier:  Sino Biological 11073H03H100
Encompass_Preferred
A DNA sequence encoding the human NTRK1 (NP_002520.2) extracellular domain (Met 1-Pro 382) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-6466