Learn More
Sino Biological Recombinant Human TrkA Protein (aa 194-413, His Tag), HPLC-verified
Supplier:  Sino Biological 11073H07H2100
Encompass_Preferred
A DNA sequence encoding the amino acid sequence (Pro 194-Glu 413) of human NTRK1 (NP_002520.2), corresponding to the Ig-like C2-type 1 & 2 domains, was expressed and purified, with a N-terminal polyhistidine tag.
Catalog No. 50-161-6470