Learn More
Sino Biological Recombinant Human TrkA Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11073H08H100
Encompass_Preferred
A DNA sequence encoding the human NTRK1 (NP_002520.2) extracellular domain (Met 1-Pro 382) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-6472