Learn More
Sino Biological Recombinant Human VLDL Receptor/VLDLR Protein (His Tag)
Supplier:  Sino Biological 11075H08H100
Encompass_Preferred
A DNA sequence encoding the human VLDLR isoform alpha (NP_003374.3) extracellular domain (Met 1-Ser 797) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6476