Learn More
Sino Biological Recombinant Human CEACAM-5/CD66e Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11077H08H100
Encompass_Preferred
A DNA sequence encoding the mature form of human CEACAM5 (NP_004354.2) (Met 1-Ala 685) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6482