Learn More
Sino Biological Recombinant Human Glucokinase Protein
Catalog No. 501616485
Encompass_Preferred
A DNA sequence encoding the human glucokinase isoform 1 (NP_000153.1) (Leu 2-Gln 465) was expressed, fused with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-6485 Supplier Sino Biological Supplier No. 11078HNCE50
Add to Cart
Add to Cart
A DNA sequence encoding the human glucokinase isoform 1 (NP_000153.1) (Leu 2-Gln 465) was expressed, fused with two additional amino acids (Gly & Pro) at the N-terminus.