Learn More
Sino Biological Influenza A H7N7 (A/Netherlands/219/2003) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 11082V08B100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of Influenza A virus (A/Netherlands/219/2003 (H7N7)) (AAR02640.1) hemagglutinin (Met 8-Phe 544) (Native, HA1+HA2) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6495