Learn More
Sino Biological Recombinant Human R-Spondin 1/RSPO1 Protein, HPLC-verified
Supplier:  Sino Biological 11083HNAS100
Encompass_Preferred
A DNA sequence encoding the human RSPO1 (NP_001033722.1) (Met1-Ala263) was expressed and purified.
Catalog No. 50-161-6506