Learn More
Sino Biological Recombinant Human Insulin Receptor Protein (ECD, short isoform, His Tag), HPLC-verified
Supplier:  Sino Biological 11086H08H100
Encompass_Preferred
A DNA sequence encoding the human INSR isoform short (NP_001073285.1) extracellular domain (Met 1-Lys 944) was expressed fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6517