Learn More
Sino Biological Recombinant Rhesus DKK-1 Protein (256 Asn/Gln, His Tag)
Supplier:  Sino Biological 11089K08H100
Encompass_Preferred
A DNA sequence encoding the rhesus DKK1 (XP_001098844.1) precursor (Met 2-His 266) (256 Asn / Gln) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-161-6533