Learn More
Sino Biological Recombinant Human CD74 Protein (ECD, His Tag)
Supplier:  Sino Biological 11091H07H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Gln 73-Met 232) of human CD74 isoform b (NP_004346.1) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6538