Learn More
Sino Biological Recombinant Human SMYD2 Protein (His Tag)
Supplier:  Sino Biological 11093H07B50
Encompass_Preferred
A DNA sequence encoding the full length of human SMYD2 (NP_064582.2) (Met 1-His 433) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6543