Learn More
Sino Biological Recombinant Human ABHD4 Protein (His Tag)
Supplier:  Sino Biological 11097H07B100
Encompass_Preferred
A DNA sequence encoding the full length of human ABHD4 (NP_071343.2) (Met 1-Asp 342) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-6550