Learn More
Chemglass Life Sciences Distilling Head, 14/20 Joint Size, 110mm Condenser Length, Approx. 105mm Height x 210mm Width
SDP
Catalog No. CG123805
Encompass_Preferred
Distilling Head, 14/20 Joint Size, 110mm Condenser Length, Approx. 105mm Height x 210mm Width. Simple distillation head with a condenser sealed to the adapter at a 75º angle. The top outer joint on the 14/20 size can be converted to a 10/18 thermometer joint by using a CG-1016-27 bushing adapter. The sidearm is sealed at a height for use with a 1” immersion thermometer. The inner drip tip joint on the condenser and inner joint on the uptake tube are the same size.
Catalog No. CG123805 Supplier Chemglass Life Sciences Supplier No. CG123805
Distilling Head, 14/20 Joint Size, 110mm Condenser Length, Approx. 105mm Height x 210mm Width. Simple distillation head with a condenser sealed to the adapter at a 75º angle. The top outer joint on the 14/20 size can be converted to a 10/18 thermometer joint by using a CG-1016-27 bushing adapter. The sidearm is sealed at a height for use with a 1” immersion thermometer. The inner drip tip joint on the condenser and inner joint on the uptake tube are the same size.