Learn More
USA Scientific Inc .1-10ul flat gel tip, .17 mm
Catalog No. 502745392
Encompass
10 uL flat sequencing gel tip, 0.17 mm OD orifice, 96 tips/rack, 2 racks/pack (192 tips)
Catalog No. 50-274-5392 Supplier USA Scientific Inc Supplier No. 10122600
10 uL flat sequencing gel tip, 0.17 mm OD orifice, 96 tips/rack, 2 racks/pack (192 tips)