Learn More
Sino Biological Recombinant Human IL-18Rα Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 11102H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human IL18R1 (NP_003846.1) (Met 1-Arg 329) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6556