Learn More
eMolecules​ 111025-46-8 | Pioglitazone | AA Blocks LLC | MFCD00865504 | 356.440 | C19H20N2O3S | 0.000 | CCc1ccc(CCOc2ccc(CC3SC(=O)NC3=O)cc2)nc1 | 1g | 721597744
Catalog No. NC3967240 Shop All eMolecules Inc. Products
Encompass_Preferred
Pioglitazone | AA Blocks LLC | 111025-46-8 | MFCD00865504 | 356.440 | C19H20N2O3S | 0.000 | CCc1ccc(CCOc2ccc(CC3SC(=O)NC3=O)cc2)nc1 | 1g | 721597744
Catalog No. NC3967240 Supplier eMolecules​ Supplier No. 721597744
Pioglitazone | AA Blocks LLC | 111025-46-8 | MFCD00865504 | 356.440 | C19H20N2O3S | 0.000 | CCc1ccc(CCOc2ccc(CC3SC(=O)NC3=O)cc2)nc1 | 1g | 721597744