Learn More
Sino Biological Recombinant Human PEDF/Serpin F1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11104H08H50
Encompass_Preferred
A DNA sequence encoding the human SERPINF1 (NP_002606.3) (Met 1-Pro 418) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6559