Learn More
Sino Biological Recombinant Human Neuroserpin Protein (His Tag)
Supplier:  Sino Biological 11107H08H50
Encompass_Preferred
A DNA sequence encoding the human SERPINI1 (NP_005016.1) (Met 1-Leu 410) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-6561