Learn More
Cayman Chemical 1113 14dIhydro15keto Pro 500ug
SDP
Catalog No. 502968429
Encompass_Preferred
A metabolite of PGD2; appears in plasma of normal males within 10 minutes of tritiated PGD2 infusion or inhalation
Catalog No. 50-296-8429 Supplier Cayman Chemical Supplier No. 16540500 UG
A metabolite of PGD2; appears in plasma of normal males within 10 minutes of tritiated PGD2 infusion or inhalation