Learn More
Cayman Chemical 1113 14dIhydro15keto Pro 50ug
SDP
Supplier:  Cayman Chemical 1878750 UG
Encompass_Preferred
An internal standard for the quantification of 11β-13,14-dihydro-15-keto PGF2α by GC- or LC-MS
Catalog No. 50-296-7811