Learn More
Cayman Chemical 1113 14dIhydro15keto Pro 50ug
SDP
Catalog No. 502966632
Encompass_Preferred
A metabolite of PGD2; appears in plasma of normal males within 10 minutes of tritiated PGD2 infusion or inhalation
Catalog No. 50-296-6632 Supplier Cayman Chemical Supplier No. 1654050 UG
A metabolite of PGD2; appears in plasma of normal males within 10 minutes of tritiated PGD2 infusion or inhalation