Learn More
Sino Biological Recombinant Human S100A12 Protein, HPLC-verified
Supplier:  Sino Biological 11143HNAE100
Encompass_Preferred
A DNA sequence encoding the native human S100A12 (NP_005612.1) (Met 1-Glu 92) was expressed.
Catalog No. 50-161-6584