Learn More
Sino Biological Recombinant Human S100A9 Protein
Supplier:  Sino Biological 11145HNAE100
Encompass_Preferred
A DNA sequence encoding the native form of human S100A9 (NP_002956.1) (Met 1-Pro 114) was expressed.
Catalog No. 50-161-6590