Learn More
Sino Biological Recombinant Human Galectin-9/LGALS9 Protein (hFc Tag)
Supplier:  Sino Biological 11147H01H50
Encompass_Preferred
A DNA sequence encoding the human LGALS9 (NP_002299.2) (Ala2-Thr323) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-6595