Learn More
Sino Biological Recombinant Human CD69 Protein (His Tag)
Supplier:  Sino Biological 11150H08H50
Encompass_Preferred
A DNA sequence encoding the human CD69 (NP_001772.1) extracellular domain (Ser 62-Lys 199) was fused with a signal peptide at the N-terminus and a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6599