Learn More
Sino Biological Recombinant Human Neuroligin 3/NLGN3 Protein (His Tag)
Supplier:  Sino Biological 11160H08H100
Encompass_Preferred
A DNA sequence encoding the human NLGN3 isoform 2 (Q9NZ94-2) extracellular domain (Met 1-Ser 689) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6613