Learn More
Sino Biological Recombinant Human Annexin VI/ANXA6 Protein (His Tag)
Supplier:  Sino Biological 11161H08E100
Encompass_Preferred
A DNA sequence encoding the human ANXA6 / Annexin A6 / Annexin6 (P08133) (Met 1-Asp 673) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6615