Learn More
Sino Biological Recombinant Human CADM1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 11168H08H100
Encompass_Preferred
A DNA sequence encoding the human CADM1 (NP_055148.3) extracellular domain (Met 1-His 374) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6621