Learn More
Sino Biological Recombinant Human PLTP Protein (His Tag)
Supplier:  Sino Biological 11171H08H50
Encompass_Preferred
A DNA sequence encoding the human PLTP isoform 1 (P55058-1) (Met 1-Val 493) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-6626